missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Factor H-related 4/CFHR4 Antibody (640212) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5980JF525
This item is not returnable.
View return policy
Description
Complement Factor H-related 4/CFHR4 Monoclonal antibody specifically detects Complement Factor H-related 4/CFHR4 in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| Complement Factor H-related 4/CFHR4 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| complement factor H-related protein 4, CFHL4, complement factor H-related 4, FHR4, FHR-4 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Complement Factor H-related 4/CFHR4, Glu20-Glu331, Accession # Q92496 | |
| RUO | |
| 10877 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 640212 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction