Learn More
Complement Component C1qA, Mouse anti-Human, Clone: C1QA/2953, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79782-100UG
3816.50 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Descrizione
Complement Component C1qA Monoclonal specifically detects Complement Component C1qA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifica
| Complement Component C1qA | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P02745, P02745 | |
| C1QA | |
| Recombinant fragment (around aa 104-237) of human Complement Component C1qA protein (exact sequence is proprietary) | |
| 100 μg | |
| Immunology, Innate Immunity | |
| 712 | |
| Store at 4°C. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| C1QA/2953 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| C1QA complement component 1, q subcomponent, A chain | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto