Learn More
PUMP SUGFUNKTION
Brand: Fisherbrand
PUMP SUGFUNKTION
Additional Details : Weight : 2.85000kg
Description
- 15 mL/s aspiration rate of liquid
- 1.8m vacuum hose included
- 1.2L PP autoclavable receiver jar with automatic flow stop
- 2 in-line hydrophobic filters
- Vacuum regulator
- Dial vacuum gauge
- Lightweight, compact, portable, standard-duty operation
- Not recommended for organic, basic and acidic vapours
Applications: Aspiration and Vacuum filtration