Learn More
PUMP SUGFUNKTION
Brand:  Fisherbrand
PUMP SUGFUNKTION
Additional Details : Weight : 2.85000kg
Product Code. 11532763
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • 15 mL/s aspiration rate of liquid
  • 1.8m vacuum hose included
  • 1.2L PP autoclavable receiver jar with automatic flow stop
  • 2 in-line hydrophobic filters
  • Vacuum regulator
  • Dial vacuum gauge
  • Lightweight, compact, portable, standard-duty operation
  • Not recommended for organic, basic and acidic vapours
Applications: Aspiration and Vacuum filtration