Learn More
Color Coded pH Buffer Solution Tested at 20°C, Reagecon™
Use color coded pH buffers for ease of use. Reagecon™ Color Coded pH Buffer Solutions Tested at 20°C are certified and NIST traceable.
204.00 NOK - 330.00 NOK
Products 6
Product Code Brand Quantity Color Price Quantity & Availability  
Product Code Brand Quantity Color Price Quantity & Availability  
11486424
View Documents Promotion Details
Reagecon
11005C
500 mL Blue
212.00 NOK
500mL
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
12808861
View Documents Promotion Details
Reagecon
10405C
500 mL Red
204.00 NOK
500mL
View alternative products
Save Up to 
»
12201473
View Documents Promotion Details
Reagecon
10705C
500 mL Yellow
204.00 NOK
500mL
View alternative products
Save Up to 
»
Estimated Shipment: 21-05-2024
to see stock.
11404579
View Documents Promotion Details
Reagecon
1100C
1 L Blue
317.00 NOK
1 L
View alternative products
Save Up to 
»
Estimated Shipment: 21-05-2024
to see stock.
11464569
View Documents Promotion Details
Reagecon
1040C
1 L Red
300.00 NOK
1 L
View alternative products
Save Up to 
»
Estimated Shipment: 21-05-2024
to see stock.
11484569
View Documents Promotion Details
Reagecon
1070C
1 L Yellow
330.00 NOK
1 L
View alternative products
Save Up to 
»
Estimated Shipment: 21-05-2024
Description
  • Colored pH Buffer Solutions
  • Tested at 20°C 
  • Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
  • NIST traceable
  • Presented in various pack sizes