Learn More
STEMCELL Technologies Collagenase Type III
For digestion of native collagen fibrils
Brand: STEMCELL Technologies 07422
Additional Details : Weight : 0.00010kg
Description
- Collagenase is a protease consisting of a single polypeptide chain approximately 1,000 amino acid residues in length
- Collagenase is capable of digesting native collagen fibrils commonly found in connective tissues and therefore is frequently used for tissue dissociation
- Collagenase preparations contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler & Yaron)
- Collagenase Type III contains low levels of proteolytic activity, especially for caseinase, clostripain, and trypsin
- Collagenase Type III has been used for the digestion of mammary glands (Rønnov-Jessen et al.), colon cancer stem cells (Prince et al.), and breast epithelial tumor cells (Chan et al.)
Specifications
Digestion of Native Collagen Fibrils | |
100 mg |