missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor VII Antibody (RFF-VII/1) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-45150JF525
This item is not returnable.
View return policy
Description
Coagulation Factor VII Monoclonal antibody specifically detects Coagulation Factor VII in Human samples. It is validated for ELISA,Western Blot,Functional Assay
Specifications
| Coagulation Factor VII | |
| Monoclonal | |
| Janelia Fluor 525 | |
| coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA | |
| Purified human Coagulation Factor VII | |
| 0.25 mL | |
| Cardiovascular Biology | |
| 2155 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Western Blot, Functional Assay | |
| RFF-VII/1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction