Learn More
Abnova™ CLTA (Human) Recombinant Protein
Human CLTA full-length ORF ( AAH19287, 1 a.a. - 218 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001211-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAELDPFGAPAGAPGGPALGNGVAGAGEEDPAAAFLAQQESEIAGIENDEAFAILDGGAPGPQPHGEPPGGPDAVDGVMNGEYYQESNGPTDSYAAISQVDRLQSEPESIRKWREEQMERLEALDANSRKQEAEWKEKAIKELEEWYARQDEQLQKTKANNRAAEEAFVNDIDESSPGTEWERVARLCDFNPKSSKQAKDVSRMRSVLISLKQAPLVH
Specifications
AAH19287 | |
clathrin, light chain (Lca) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLTA | |
GST |
1211 | |
Wheat germ expression system | |
25 μg | |
LCA | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |