Learn More
Nickel Electro™ Cooling Coil
Nickel Electro™ Cooling Coil immerses into Clifton™ Stirred or Circulating water bath, behind the Stirrer, without affecting bath working area. When connected to a cold water supply it provides counter cooling depending on tap water temperature and flow rate.
Brand: Nickel Electro™ NE4-CC
Additional Details : Weight : 1.00000kg
Description
- This Cooling Coil does not provide forced cooling as provided with DC range of Immersion Dip Coolers.
- A Lid is recommended in use.
- Stainless steel
- For use with cold running water supply.
Specifications
Cooling Coil | |
20 x 10 x 10 cm | |
20 x 10 x 10 cm |