Learn More
Fisherbrand™ Clear Graduated Glass Bottles with Black Phenolic Caps
Click to view available options
Capacity (English):
2 oz.
8 oz.
Capacity (Metric):
60 mL
240 mL
Description
- Graduated in both milliliters and ounces
- Clear glass bottles are designed with a slight shoulder for easy pouring and handling
- Wide mouth facilitates easy mixing, handling, and transfer of contents
Specifications
Closure Liner | Polyethylene Cone |
Closure Material | Phenolic |
Capacity (Metric) | 60 mL |
Capacity (English) | 2 oz. |
Color | Clear |
Material | Glass (Type 3) |
Quantity | 48 |
Closure Color | Black |
Neck Finish | 38-400 |
Closure Included | Yes |