missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CKS2 (Human) Recombinant Protein
Human CKS2 full-length ORF ( AAH06458, 1 a.a. - 79 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001164-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAHKQIYYSDKYFDEHYEYRHVMLPRELSKQVPKTHLMSEEEWRRLGVQQSLGWVHYMIHEPEPHILLFRRPLPKDQQK
Specifications
AAH06458 | |
CDC28 protein kinase regulatory subunit 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CKS2 | |
GST |
1164 | |
Wheat germ expression system | |
25 μg | |
CKSHS2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |