Learn More
cis-Dibromocypermethric acid 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30318809
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 30318809
Brand: Dr. Ehrenstorfer DREA12244000AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material cis-Dibromocypermethric acid
CAS 63597-73-9
Analyte or Component Names cis-Dibromocypermethric acid
Shipping Condition +20°C
Molecular Formula C8 H10 Br2 O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Cyclopropanecarboxylic acid, 3-(2,2-dibromoethenyl)-2,2-dimethyl-, cis-, rel-(1R,3R)-3-(2,2-Dibromoethenyl)-2,2-dimethylcyclopropanecarboxylic acid, (±)-Dibromo-cis-cypermethric acid, cis-(±)-Dibromocypermethric acid, cis-2,2-Dimethyl-3-(2,2-dibromovinyl)cyclopropanecarboxylic acid, cis-3-(2,2-Dibromovinyl)-2,2-dimethylcyclopropane-1-carboxylic acid, cis-DL-3-(2,2-Dibromovinyl)-2,2-dimethylcyclopropanecarboxylic acid, cis-Decamethrinic acid, cis-DBCA
InChI Formula InChI=1S/C8H10Br2O2/c1-8(2)4(3-5(9)10)6(8)7(11)12/h3-4,6H,1-2H3,(H,11,12)/t4-,6-/m0/s1
SMILES CC1(C)[C@@H](C=C(Br)Br)[C@H]1C(=O)O
IUPAC Name (1R,3R)-3-(2,2-dibromoethenyl)-2,2-dimethylcyclopropane-1-carboxylic acid
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 297.97
Quantity 1 mL
Formula Weight 295.9048 g/mol