Learn More
Macherey-Nagel™ Chromabond™ Sioh Polypropylene Column BIG Pack
Brand:  Macherey-Nagel™ 730107.250
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12476546
Quantity Price
1 12040.00 NOK / Pack of 250
3 11322.42 NOK / Pack of 250
Estimated Shipment: 04-11-2021
Specifications
SPE Column
6 mL
2000mg
SPE
Chromabond Sioh
45 µm
Silica
60 Å
500 m2/g
Solid Phase Extraction