missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromabond™ Alox B Polypropylene Column

Brand:  Macherey-Nagel™ 730020

Additional Details : Weight : 0.01000kg

Product Code. 10756512

  • 1950.00 NOK / Pack of 30

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

6 mL
Solid Phase Extraction
Chromabond Alox B
150 m2/g
SPE
60 to 150 μm
Solid Phase Extraction Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers