Learn More
chlorodimethylethylsilane, 97%, Thermo Scientific™
Catalog Number p-4228506
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
6917-76-6
C4H11ClSi
122.67
MFCD00009918
AVDUEHWPPXIAEB-UHFFFAOYSA-N
ethyldimethylchlorosilane, chlorodimethylethylsilane, chloro ethyl dimethylsilane, dimethylethylchlorosilane, dimethylethylsilyl chloride, ethyldimethylsilyl chloride, chloro ethyl dimethylsilane #, silane, chlorodimethylethyl-,
138863
chloro-ethyl-dimethylsilane
CC[Si](C)(C)Cl
6917-76-6
C4H11ClSi
122.67
MFCD00009918
AVDUEHWPPXIAEB-UHFFFAOYSA-N
ethyldimethylchlorosilane, chlorodimethylethylsilane, chloro ethyl dimethylsilane, dimethylethylchlorosilane, dimethylethylsilyl chloride, ethyldimethylsilyl chloride, chloro ethyl dimethylsilane #, silane, chlorodimethylethyl-,
138863
chloro-ethyl-dimethylsilane
CC[Si](C)(C)Cl
Color Colorless
Density 0.8730g/mL
Boiling Point 89.0°C to 91.0°C
Flash Point −8°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4050 to 1.4070 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.873
Formula Weight 122.67
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material chlorodimethylethylsilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chlorodimethylethylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.