Learn More
Chloroacetyl isocyanate, 90%, Tech., Thermo Scientific Chemicals
Catalog Number p-85918
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
4461-30-7
C3H2ClNO2
119.51
MFCD00002034
MOVMEFHWBOWMFU-UHFFFAOYSA-N
chloroacetyl isocyanate, acetyl isocyanate, chloro, unii-h5wf2amh7j, chloroacetyl isocyante, h5wf2amh7j, snxhcfhai zyp@, chloroacetylisocyanate, acmc-20alh0, chloro-acetyl isocyanate, alpha-chloroacetyl isocyanate
78209
2-chloroacetyl isocyanate
C(C(=O)N=C=O)Cl
4461-30-7
C3H2ClNO2
119.51
MFCD00002034
MOVMEFHWBOWMFU-UHFFFAOYSA-N
chloroacetyl isocyanate, acetyl isocyanate, chloro, unii-h5wf2amh7j, chloroacetyl isocyante, h5wf2amh7j, snxhcfhai zyp@, chloroacetylisocyanate, acmc-20alh0, chloro-acetyl isocyanate, alpha-chloroacetyl isocyanate
78209
2-chloroacetyl isocyanate
C(C(=O)N=C=O)Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Yellow
Density 1.4030g/mL
Boiling Point 50.0°C to 55.0°C (20.0 mmHg)
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula ClCH2CONCO
Quantity 1 g
Merck Index 15, 2117
Specific Gravity 1.403
Formula Weight 119.51
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Chloroacetyl isocyanate, tech., 90%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.