Learn More
Chloroacetonitrile, 98+%, Thermo Scientific Chemicals
Catalog Number p-85888
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
107-14-2
C2H2ClN
75.5
MFCD00001885
RENMDAKOXSCIGH-UHFFFAOYSA-N
chloroacetonitrile, chloracetonitrile, acetonitrile, chloro, chloromethyl cyanide, monochloroacetonitrile, chloroethanenitrile, monochloromethyl cyanide, usaf kf-5, alpha-chloroacetonitrile, acetonitrile, 2-chloro
7856
CHEBI:82416
2-chloroacetonitrile
C(C#N)Cl
107-14-2
C2H2ClN
75.5
MFCD00001885
RENMDAKOXSCIGH-UHFFFAOYSA-N
chloroacetonitrile, chloracetonitrile, acetonitrile, chloro, chloromethyl cyanide, monochloroacetonitrile, chloroethanenitrile, monochloromethyl cyanide, usaf kf-5, alpha-chloroacetonitrile, acetonitrile, 2-chloro
7856
CHEBI:82416
2-chloroacetonitrile
C(C#N)Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.1890g/mL
Boiling Point 124.0°C to 126.0°C
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CN
Refractive Index 1.4200 to 1.4240
Quantity 100 mL
Beilstein 02,201
Fieser 01,129; 06,102
Specific Gravity 1.189
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol, diethyl ether, hydrocarbons
Formula Weight 75.5
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Chloroacetonitrile, 98+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chloroacetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.