Learn More
Lovibond™ Chlorine pH test kit AF 116B
AF 116B Comparator 2000+ chlorine pH test kit measures chlorine 0.2 - 4.0 mg/L pH 6.8 - 8.4 includes test discs 3/40B and 2/1J accessories tablets and carry case
Brand: Lovibond™ 411160
Additional Details : Weight : 1.31700kg
Description
The Comparator 2000+ is a versatile colorimetric system for water analysis. Comfortable handling, no compromise on accuracy and reproducibility.
Usable in different applications
The Comparator 2000+ is versatile: it is used in swimming pools as well as in research centers, universities or general drinking water treatment.
Daylight lamp
So that you can always measure safely and stably under unfavorable visibility conditions, we recommend the use of the (optional) mains or battery-powered daylight lamp. So the lighting conditions always remain the same. No matter where you are measuring.
Various analyses possible
For the Comparator 2000+, more than 400 color discs and numerous reagents are available. Your combination is guaranteed. And if you have to change them once, it works without opening the comparator.
Integrated prism
The integrated prism visually brings the glass standards of the color disc and the colored sample into view.
Specifications
Chlorine and pH | |
chlorine 0.2 to 4.0 mg/L pH 6.8 to 8.4 | |
Test Kit |
Safety and Handling
- AF116 PH AND CHLORINE KIT
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Specific target organ toxicity Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H372-Causes damage to organs.
Precautionary Statement
- P233-Keep container tightly closed.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P314-Get medical advice/attention if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : potassium bromide, butan-2-ol, potassiumiodide