Learn More
Saint Gobain Life Sciences Chemware™ PFA Graduated Wide Mouth Wash Bottle
Click to view available options
Capacity:
100 mL (3.4 oz.)
Description
- Wide-mouth for easy filling
- Chemically inert and leak-proof
- Flexible nozzle with 360° swivel; small orifice allows accurate content dispensing
- Transparent and break resistant
- Molded-in graduation marks
Specifications
Capacity | 100 mL (3.4 oz.) |
Height (English) | 6.3 in. |
Height (Metric) | 159.5 mm |
Product Type | Wash Bottle |
Capacity (Metric) | 100 mL |
Capacity (English) | 3.4 oz. |
Color | Transparent |
Material | PFA |
Neck ID (Metric) | 25.5 mm |
Graduated | Yes |