Learn More
Saint Gobain Life Sciences Chemware™ PFA Narrow Neck Graduated Bottle
Catalog Number p-8369101
Click to view available options
Capacity:
100 mL (3.4 oz.)
250 mL (8.5 oz.)
5000 mL (1.3 gal.)
Store virtually all corrosive chemicals, including hydrofluoric, nitric, and perchloric acids. Chemware™ PFA Narrow Neck Graduated Bottle is chemically inert and heat resistant with a non-stick surface that is ideal for environmental sampling.
  • Ultra-pure - the lowest metal ion content of all fluoropolymers
  • Unbreakable
  • Chemically inert
  • Non-stick, easy to clean
  • Use for storage in pharmaceutical and semiconductor processing (intermediate stages)
Capacity 100 mL (3.4 oz.)
Product Type Bottle
Capacity (Metric) 100 mL
Capacity (English) 3.4 oz.
Diameter (Metric) Inner Neck 16.7 mm
Height (Metric) With Closure 126 mm
Material PFA
Graduated Yes
Mouth Narrow