Learn More
Merck Millipore Chemicon™ In Vitro Vascular Permeability Assay
For evaluating effects of chemicals and drug compounds on endothelial cell adsorption, transport and permeability
Brand: Merck Millipore ECM644
Additional Details : Weight : 0.50000kg
Specifications
In Vitro Vascular Permeability Assay | |
Evaluating effects of chemicals and drug compounds on endothelial cell adsorption, transport and permeability | |
24 assays |
Fluorescent | |
Black plate, 24-well opaque/permeability insert assembly collagen-coated, receiver tray, FITC-dextran solution, cell stain, forceps | |
Store kit materials at 2-8°C; use within 4 months from date of receipt |