Learn More
Merck Millipore Chemicon™ In Vitro Vascular Permeability Assay
For evaluating effects of chemicals and drug compounds on endothelial cell adsorption, transport and permeability
Brand:  Merck Millipore ECM644
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 11392940
  • 8233.00 NOK / Each
Estimated Shipment
05-03-2021
Specifications
In Vitro Vascular Permeability Assay
Evaluating effects of chemicals and drug compounds on endothelial cell adsorption, transport and permeability
24 assays
Fluorescent
Black plate, 24-well opaque/permeability insert assembly collagen-coated, receiver tray, FITC-dextran solution, cell stain, forceps
Store kit materials at 2-8°C; use within 4 months from date of receipt