Learn More
Unity Lab Services™ Chemical Filtering Hood IQOQ Service
Product Code. 17917792
Change view
Click to view available options
For Use With (Equipment):
Chemical Filtering Hood
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17917792 Chemical Filtering Hood 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17917792 Supplier Unity Lab Services™ Supplier No. IOQCHEMVS
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
IQOQ service for chemical filtering hoods.
  • IQ:Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order.
  • Verifies equipment and any other accessories are assembled and installed. Any abnormal event(s) observed during the IQ is documented in the IQ protocol.
  • Verifies that the installation site and equipment environment meet manufacturer-specified environmental requirements.
  • Configuration information for each system component gets recorded.
  • Installation of Equipment is NOT Included.
  • OQ: System component information detailed configuration information for each system component have been recorded (as applicable).
  • Verifies important equipment functions and ensures that the equipment operates as expected by the manufacturer and in conformance to standards and requirements.
  • Key test varies based on relevant certification standard (NSF49/EN12469/DIN12480). Examples: filter leakage, DEHS, air velocity, electrical safety, air-flow smoke pattern, KI-DISCUS™.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Chemical Filtering Hood
Product Type IQ/OQ Service