missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD4 Antibody [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18483JF669
This item is not returnable.
View return policy
Description
CHCHD4 Polyclonal antibody specifically detects CHCHD4 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| CHCHD4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 131474 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 669 | |
| CHDH4, coiled-coil-helix-coiled-coil-helix domain containing 4, Coiled-coil-helix-coiled-coil-helix domain-containing protein 4, FLJ31709, MIA40mitochondrial intermembrane space import and assembly protein 40, mitochondrial intermembrane space import and assembly 40 homolog, TIMM40, translocase of inner mitochondrial membrane 40 homolog | |
| Recognizes region between residues Between between 92 and 142.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction