Learn More
Implen™ Certified Holmium Glass and Neutral Density Glass Filter Set for the control of wavelength and photometric accuracy of the NanoPhotometer™ NP80 and C40
Certified Holmium Glass and Neutral Density Glass Filter Set for the control of wavelength and photometric accuracy of the NanoPhotometer™ NP80 and C40 (cuvette)
Brand: Implen™ N-4080-HNF
This item is not returnable.
View return policy
Description
The certified holmium and neutral density glass filter are an optional accessory used as a control of wavelength and photometric accuracy for the NanoPhotometer™ Cuvette applications.Specifications
NanoPhotometer C40/NP80 | |
Holmium and Neutral Density Glass Filter Set | |
Reusable | |
NanoPhotometer C40/NP80 | |
Holmium and Neutral Glass |
10 mm | |
English | |
Wavescan | |
Holmium and Neutral Density Glass Filter Set |