Learn More
Hettich™ Centrifuge Adapters for 8-place Angel Rotors
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
375.00 NOK
Specifications
Type | Adapter |
---|---|
For Use With (Equipment) | ROTOFIX 32 A and ROTINA 380/380 R Centrifuges with Rotors no. 1418, 1721 |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
10004467
|
Hettich™
1467 |
4 x 1.1 to 15 mL Tubes |
375.00 NOK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10412305
|
Hettich™
1468 |
50 mL |
375.00 NOK
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Centrifuge Adapters for 8-place Angle Rotors
- For use with ROTOFIX 32 A and ROTINA 380/380 R Centrifuges
- For Rotors no. 1418, 1721
Specifications
Adapter | |
ROTOFIX 32 A and ROTINA 380/380 R Centrifuges with Rotors no. 1418, 1721 |