Learn More
CDK4 Mouse anti-Human, Unconjugated, Clone: 1G2C12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66950-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The CDK4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDK4. This antibody recognizes human, mouse, rat antigen. The CDK4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
CDK4 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry | |
1G2C12 | |
Unconjugated | |
20 μL | |
1019 | |
IgG1 |