missing translation for 'onlineSavingsMsg'
Learn More

CD95 (APO-1/Fas) Mouse anti-Canine, Human, eFluor® 450, Clone: DX2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0959-41

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15589696

  • 1034.00 NOK / 25 tests


Estimated Shipment
08-09-2020
Add to basket

Description

Description

CD95 (Fas, APO-1), a 46 kDa transmembrane glycoprotein, is a cell death receptor of the TNFR superfamily. Stimulation of CD95 results in aggregation of its intracellular death domains, formation of the death-inducing signaling complex (DISC) and activation of caspases. In type I cells caspase 3 is activated by high amounts of caspase 8 generated at the DISC, in type II cells low concentration of caspase 8 activates pathway leading to the release of cytochrome c from mitochondria and activation of caspase 3 by cytochom c. Besides its roles in induction of apoptosis, Fas also triggers pro-inflammatory cytokine responses.
Specifications

Specifications

CD95 (APO-1/Fas)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P25445
ALPS1A, APO-1, APT1, Apo-1 Fas, CD95, FAS1, FASTM, TNFRSF6
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Canine, Human
Flow Cytometry
DX2
eFluor™ 450
FAS
Liquid
FAS
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
355, 486469
Videos
Documents

For Research Use Only.