missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD93 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550583
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total CD93 / C1qR protein.
CD93 Polyclonal antibody specifically detects CD93 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CD93 | |
Polyclonal | |
Unconjugated | |
Cd93 | |
6030404G09Rik; AA145088; Aa4; AA4.1; AW555904; C1q receptor; C1q receptor 1; C1q/MBL/SPA receptor; C1qR; C1qR(P); C1qr1; C1qRP; Cd93; CD93 antigen; CD93 molecule; CDw93; Cell surface antigen AA4; complement component 1 q subcomponent receptor 1; complement component 1, q subcomponent, receptor 1; complement component C1q receptor; dJ737E23.1; ECSM3; Ly68; ly-68; Lymphocyte antigen 68; matrix-remodeling-associated protein 4; matrix-remodelling associated 4; MXRA4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22918 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9NPY3 | |
Cd93 | |
Synthetic peptide corresponding to a region derived from internal residues of human CD93 molecule | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |