missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90 Monoclonal Antibody (G7), PE, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen A14726
Additional Details : Weight : 0.01000kg
Description
CD90 Monoclonal antibody specifically detects CD90 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD90 | |
Monoclonal | |
0.1 mg/mL | |
PBS with <0.1% sodium azide | |
P01831 | |
Thy1 | |
MST cell hybridoma C6/G8 from Mouse T cell hybridoma C6/G8 | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2c κ |
Flow Cytometry | |
G7 | |
PE | |
Thy1 | |
CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen | |
Rat | |
purified | |
RUO | |
21838 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |