missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD86 Monoclonal Antibody (GL1), PE-Cyanine7, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen A15412
Additional Details : Weight : 0.00280kg
Description
CD86 Monoclonal antibody specifically detects CD86 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD86, CD86 (B7-2) | |
Monoclonal | |
0.1 mg/mL | |
PBS with proprietary stabilizer and <0.1% sodium azide | |
P42082 | |
CD86 | |
Mouse CD86 from LPS-activated CBA/Ca mouse spleen B cells | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
GL1 | |
PE-Cyanine7 | |
CD86 | |
activation B7-2 antigen; B lymphocyte activation antigen B72; B7; B7.2; B70; B7-2; B-lymphocyte activation antigen B7-2; BU63; CD28 antigen ligand 2; Cd28l2; CD28LG2; CD86; CD86 antigen; CD86 antigen (CD28 antigen ligand 2, B7-2 antigen); CD86 antigen precursor; CD86 molecule; CLS1; CTLA-4 counter-receptor B7.2; early T cell costimulatory molecule-1; early T-cell costimulatory molecule 1; Early T-cell co-stimulatory molecule 1; ETC-1; FUN-1; LAB72; Ly58; Ly-58; MB7; MB7-2; membrane glycoprotein; MGC34413; T-lymphocyte activation antigen CD86; TS/A-2 | |
Rat | |
purified | |
RUO | |
12524 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |