missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Mouse anti-Human, Clone: CD5-5D7, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD0500
Additional Details : Weight : 0.50000kg
Description
CD5 is a 67 kDa human T-lymphocyte single-chain transmembrane glycoprotein. CD5 is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. CD5 also reacts with a subpopulation of activated B-cells and may act as a receptor in regulating T-cell proliferation. CD5 is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Diseases associated with CD5 dysfunction include thymus cancer and Richter’s Syndrome.Specifications
CD5 | |
Monoclonal | |
Unconjugated | |
CD5 | |
CD5 antigen p56-62, T-cell surface glycoprotein CD5, lymphocyte antigen T1/Leu-1 | |
Mouse | |
Purified | |
ASR | |
921 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
CD5-5D7 | |
PBS with 0.1% sodium azide | |
P06127 | |
CD5 | |
Human CD5 | |
0.5 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |