Learn More
CD42d Monoclonal Antibody (1C2), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Affymetrix eBioscience 53-0421-82
Additional Details : Weight : 0.01000kg
Description
Description: The monoclonal antibody 1C2 recognizes mouse and rat CD42d, a single transmembrane protein also known as platelet glycoprotein V (GPV). CD42d interacts with CD42a, CD42b and CD42c to form the receptor for von willebrand factor and thrombin. This functional complex plays a role in platelet adhesion. Expression of CD42d (unlike other markers such as CD41) is restricted to resting platelets and megakaryocytes and therefore is a good marker for development of megakaryocytes. Applications Reported: This 1C2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1C2 antibody has been tested by flow cytometric analysis of mouse platelets. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser
Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.Specifications
CD42d | |
Monoclonal | |
0.5 mg/ml | |
PBS with 0.09% sodium azide; pH 7.2 | |
O08770 | |
Gp5 | |
Affinity chromatography | |
RUO | |
14729, 25259 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
1C2 | |
Alexa Fluor 488 | |
Gp5 | |
Gp5; GPV; PLGPV | |
Armenian Hamster | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |