missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD40/TNFRSF5 Antibody (1C10) [Janelia Fluor™ 585], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB440JF585
This item is not returnable.
View return policy
Description
CD40/TNFRSF5 Monoclonal antibody specifically detects CD40/TNFRSF5 in Mouse samples. It is validated for Flow Cytometry,Immunoprecipitation,Agonist Activity,CyTOF
Specifications
| CD40/TNFRSF5 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5 | |
| COS-7 African green monkey SV40 transformed kidney fibroblast-like cell line-derived recombinant mouse CD40/TNFRSF5, Extracellular domain | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunoprecipitation, Agonist Activity, CyTOF | |
| 1C10 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Asthma, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Signal Transduction, Stem Cell Markers | |
| 958 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction