Learn More
CD321 (F11R) Monoclonal Antibody (CSTEM27), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1114.00 NOK - 2810.00 NOK
Specifications
Antigen | CD321 (F11R) |
---|---|
Clone | CSTEM27 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry, Immunocytochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15845463
|
Affymetrix eBioscience
53-3219-42 |
100 Tests |
2810.00 NOK
Pack of 100 |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15835463
|
Invitrogen
53-3219-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This monoclonal antibody CSTEM27 reacts with human CD321 (F11R, JAM-1, JAM-A). Applications Reported: This CSTEM27 antibody has been reported for use in immunocytochemical staining and flow cytometric analysis. This CSTEM27 antibody has been tested by immunocytochemistry of adherent cells. This can be used at less than or equal to 5 μg/mL. This CSTEM27 antibody has also been tested by flow cytometric analysis of human iPSCs. This may be used at 0.25 μg/test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD321 (F11R) Monoclonal antibody specifically detects CD321 (F11R) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
CD321 (F11R) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q9Y624 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
CSTEM27 | |
Flow Cytometry, Immunocytochemistry | |
Alexa Fluor 488 | |
Mouse | |
Human | |
50848 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |