missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300e Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA568389
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Human samples.
CD300e Polyclonal antibody specifically detects CD300e in Human, Mouse samples. It is validated for Western Blot.Specifications
CD300e | |
Polyclonal | |
Unconjugated | |
CD300E | |
BC034097; CD300 antigen like family member E; CD300 antigen-like family member E; CD300e; CD300e antigen; CD300e molecule; Cd300le; CLM2; CLM-2; CMRF35A5; CMRF35-A5; CMRF35-like molecule 2; CMRF-35-like molecule-1; Immune receptor expressed on myeloid cells 2; IREM2; IREM-2; PIgR2; PIgR-2; poly-Ig receptor 2; polymeric immunoglobulin receptor 2; TREM5 | |
Rabbit | |
affinity chromatography | |
RUO | |
217306, 342510 | |
-20°C | |
Liquid |
Western Blot | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q496F6, Q8K249 | |
CD300E | |
Synthetic protein corresponding to residues near the C terminal of human CD300e. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |