Learn More
CD138 (Syndecan-1) Monoclonal Antibody (DL-101), Super Bright™ 600, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1450.00 NOK - 3660.00 NOK
Specifications
Antigen | CD138 (Syndecan-1) |
---|---|
Clone | DL-101 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15712496
|
Affymetrix eBioscience
63-1389-42 |
100 Tests |
3660.00 NOK
Pack of 100 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15702496
|
Affymetrix eBioscience
63-1389-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The DL-101 monoclonal antibody reacts with human CD138, also known as syndecan-1. CD138 is a transmembrane protein containing chondroitin sulfate and heparin sulfate moieties responsible for binding to the extracellular matrix components. CD138 is not expressed by mature B cells but is present on pre-B cells and the finally differentiated plasma cells. This DL-101 antibody has been pre-titrated and tested by flow cytometric analysis of the U266 cell line. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 600 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 600 nm. We recommend using a 610/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product No. SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat.
CD138 (Syndecan-1) Monoclonal antibody specifically detects CD138 (Syndecan-1) in Human samples. It is validated for Flow Cytometry.Specifications
CD138 (Syndecan-1) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P18827 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
DL-101 | |
Flow Cytometry | |
Super Bright 600 | |
Mouse | |
Human | |
6382 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only