Learn More
CD137 Ligand (4-1BB Ligand) Monoclonal Antibody (4H3), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1246.00 NOK - 3250.00 NOK
Specifications
Antigen | CD137 Ligand (4-1BB Ligand) |
---|---|
Clone | 4H3 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15825363
|
Affymetrix eBioscience
17-5906-42 |
100 Tests |
3250.00 NOK
Pack of 100 |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15815363
|
Affymetrix eBioscience
17-5906-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This 4H3 monoclonal antibody recognizes human CD137 ligand, also known as 4-1BB ligand, TNFSF9, or CDw137L. Applications Reported: This 4H3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 4H3 antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD137 Ligand (4-1BB Ligand) Monoclonal antibody specifically detects CD137 Ligand (4-1BB Ligand) in Human samples. It is validated for Flow Cytometry.Specifications
CD137 Ligand (4-1BB Ligand) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P41273 | |
TNFSF9 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
TNFSF9 |
4H3 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
8744 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |