missing translation for 'onlineSavingsMsg'
Learn More

CD133 (Prominin-1) Rat anti-Canine, Mouse, PerCP-eFluor™ 710, Clone: 13A4, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-1331-80

Additional Details : Weight : 0.30000kg

 View more versions of this product

Product Code. 15591106

  • 1310.00 NOK / 25µg


Estimated Shipment
14-08-2020
Add to basket

Description

Description

Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. The CD133 (also named AC133) antigen is a 97 kDa glycoprotein, with 5 transmembrane domains, that shows up as a 120 kDa band on protein gels due to glycosylation. It is a cell surface marker expressed by immature hematopoietic stem cells but not mature blood cells. In addition to hematopoietic stem and progenitor cells, CD133 has been found to be a marker for other stem and progenitor cells including neural and embryonic stem cells. It has been shown to be expressed in cancers, including some leukemias and brain tumours, that may be derived from stem cells. Low level expression could also be detected in the kidney, pancreas, placenta, and fetal liver.
Specifications

Specifications

CD133 (Prominin-1)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O54990
AC133, CD133, CORD12, MCDR2, MSTP061, PROML1, RP41, STGD4
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Canine, Mouse
Flow Cytometry
13A4
PerCP-eFluor™ 710
Prom1
Liquid
PROM1
IgG1, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
19126, 488816
Videos
Documents

For Research Use Only.