missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCAM-1 Monoclonal Antibody (1.G11B1), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15439
Additional Details : Weight : 0.00280kg
Description
VCAM-1 Monoclonal antibody specifically detects VCAM-1 in Human, Porcine samples. It is validated for Flow Cytometry.Specifications
VCAM-1, CD106 (VCAM-1) | |
Monoclonal | |
0.1 mg/mL | |
PBS with <0.1% sodium azide | |
P19320 | |
VCAM1 | |
human CD106 | |
100 μg | |
Primary | |
Human, Porcine | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
1.G11B1 | |
FITC | |
VCAM1 | |
adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B | |
Mouse | |
purified | |
RUO | |
396925, 7412 | |
4° C, store in dark | |
Liquid |