missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD103 Monoclonal Antibody (LF61), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD10301
Additional Details : Weight : 0.50000kg
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility.
CD103 Monoclonal antibody specifically detects CD103 in Human samples. It is validated for Flow Cytometry.Specifications
CD103 | |
Monoclonal | |
FITC | |
Itgae | |
A530055J10; alpha-E1; alpha-M290; aM290; antigen CD103; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; CD103; DC; DCs; HML-1 antigen; human mucosal lymphocyte antigen 1, alpha polypeptide; HUMINAE; integrin alpha E; integrin alpha E, epithelial-associated; integrin alpha E1, epithelial-associated; integrin alpha M290; Integrin alpha-E; Integrin alpha-E heavy chain; Integrin alpha-E light chain; Integrin alpha-IEL; integrin subunit alpha E; integrin, alpha E; integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide); integrin, alpha E, epithelial-associated; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen | |
Mouse | |
purified | |
ASR | |
3682 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
LF61 | |
PBS with 4mg/mL BSA and 0.1% sodium azide | |
P38570 | |
Itgae | |
Human CD103 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |