missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD103 Monoclonal Antibody (2E7), FITC, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen HMCD10301
Additional Details : Weight : 0.00010kg
Description
CD103 Monoclonal antibody specifically detects CD103 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD103 | |
Monoclonal | |
0.5 mg/mL | |
PBS with <0.1% sodium azide | |
Q60677 | |
Itgae | |
Mouse CD103. from C57BL/6J mouse intestinal intraepithelial lymphocytes (IEL) | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2 |
Flow Cytometry | |
2E7 | |
FITC | |
Itgae | |
A530055J10; alpha-E1; alpha-M290; aM290; antigen CD103; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; CD103; DC; DCs; HML-1 antigen; human mucosal lymphocyte antigen 1, alpha polypeptide; HUMINAE; integrin alpha E; integrin alpha E, epithelial-associated; integrin alpha E1, epithelial-associated; integrin alpha M290; Integrin alpha-E; Integrin alpha-E heavy chain; Integrin alpha-E light chain; Integrin alpha-IEL; integrin subunit alpha E; integrin, alpha E; integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide); integrin, alpha E, epithelial-associated; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen | |
Armenian Hamster | |
purified | |
RUO | |
16407 | |
4° C, store in dark | |
Liquid |