missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR9 Antibody (882416) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8287JF525
4364.75 NOK valid until 2025-12-16
Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
CCR9 Monoclonal antibody specifically detects CCR9 in Rat samples. It is validated for Flow Cytometry,CyTOF
Specifications
| CCR9 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR-9, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28 | |
| NS0 mouse myeloma cell line transfected with rat CCR9 | |
| RUO | |
| Primary | |
| Rat | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 882416 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 10803 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction