missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL20/MIP-3 alpha Antibody (67310) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB360JF635
This item is not returnable.
View return policy
Description
CCL20/MIP-3 alpha Monoclonal antibody specifically detects CCL20/MIP-3 alpha in Human samples. It is validated for Western Blot,ELISA Standard (Matched Pair),Neutralization,ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
Specifications
| CCL20/MIP-3 alpha | |
| Monoclonal | |
| Janelia Fluor 635 | |
| beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARCLiver and activation-regulated chemokine, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38 | |
| E. coli-derived recombinant human CCL20/MIP-3 alpha , Ala27-Met96 (Asn95Asp), Accession # P78556.1 | |
| 0.1 mL | |
| Biologically Active Proteins, Chemokines and Cytokines | |
| 6364 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) | |
| 67310 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction