missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caveolin-1 Antibody (7C8) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5736JF635
This item is not returnable.
View return policy
Description
Caveolin-1 Monoclonal antibody specifically detects Caveolin-1 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry
Specifications
| Caveolin-1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085 | |
| Puified rat adipocyte low density microsomes, Accession # P41350 | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 7C8 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer, Caveolae Markers, Cellular Markers, Cholesterol Metabolism, Hypoxia, Lipid and Metabolism, Lipid Droplets, Phospho Specific, Signal Transduction | |
| 857 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction