Learn More
BINDER™ Cart with Casters
Stable Cart with Casters with locking brakes. Accessory for use with Avantgarde.Line Incubators.
Brand:  BINDER™ 9051-0018
Additional Details : Weight : 5.00000kg
Product Code. 15632206
  • 14230.29 NOK / Each
Estimated Shipment: 12-07-2024
Description
For Use With:
  • BD 23, BD 56, BD 115, BF 56, BF 115, ED 56, ED 115, FD 56, FD 115, FDL 115, FED 56, FED 115, FP 56, FP 115, KB 53, KB 115, KT 53, KT 115, KT 170, M 53, M 115
Specifications
Specifications
stable cart, casters with locking brakes, dimensions: W 1,000 x D 800 x H 780 mm
Steel
800 x 1,000 x 780 mm
BINDER Chambers BD 23, BD 56, BD 115, BF 56, BF 115, ED 56, ED 115, FD 56, FD 115, FDL 115, FED 56, FED 115, FP 56, FP 115, KB 53, KB 115, KT 53, KT 115, KT 170, M 53, M 115
Cart