missing translation for 'onlineSavingsMsg'
Learn More

Carbon felt, 1.27cm (0.5in) thick, 99.0%, Thermo Scientific Chemicals

Catalog Number p-4412843
Click to view available options
Quantity:
10 x 10 cm
30 x 30 cm
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
MFCD00133992
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
MFCD00133992

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 99.0%
Quantity 10 x 10 cm
Chemical Name or Material Carbon felt, Processed at 1400°C

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'