missing translation for 'onlineSavingsMsg'
Learn More

Carbenoxolone disodium salt, 97+%, Thermo Scientific Chemicals

Catalog Number p-7042777
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7421-40-1
C34H48Na2O7
614.73
MFCD00079043
BQENDLAVTKRQMS-SBBGFIFASA-L
carbenoxolone disodium salt, disodium 3beta-3-3-carboxylatopropanoyl oxy-11-oxoolean-12-en-30-oate, disodium 4as,6as,6br,10s,12as,12br,14br-10-3-carboxylatopropanoyl oxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,7,8,8a,10,11,12,12b,14b-dodecahydro-1h-picene-2-carboxylate
133687779
disodium;(2R,4aR,6aS,6aR,6bS,8aS,10R,12aR,14bS)-10-(3-carboxylatopropanoyloxy)-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylate
[Na+].[Na+].[H][C@@]12C[C@](C)(CC[C@]1(C)CC[C@]1(C)C2=CC(=O)[C@]2([H])[C@@]3(C)CC[C@H](OC(=O)CCC([O-])=O)C(C)(C)[C@]3([H])CC[C@@]12C)C([O-])=O
This item is not returnable. View return policy
7421-40-1
C34H48Na2O7
614.73
MFCD00079043
BQENDLAVTKRQMS-SBBGFIFASA-L
carbenoxolone disodium salt, disodium 3beta-3-3-carboxylatopropanoyl oxy-11-oxoolean-12-en-30-oate, disodium 4as,6as,6br,10s,12as,12br,14br-10-3-carboxylatopropanoyl oxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,7,8,8a,10,11,12,12b,14b-dodecahydro-1h-picene-2-carboxylate
133687779
disodium;(2R,4aR,6aS,6aR,6bS,8aS,10R,12aR,14bS)-10-(3-carboxylatopropanoyloxy)-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylate
[Na+].[Na+].[H][C@@]12C[C@](C)(CC[C@]1(C)CC[C@]1(C)C2=CC(=O)[C@]2([H])[C@@]3(C)CC[C@H](OC(=O)CCC([O-])=O)C(C)(C)[C@]3([H])CC[C@@]12C)C([O-])=O

Carbenoxolone is anti-inflammatory. It is often used as a gap junction inhibitor in neuroscience research. Carbenoxolone is also an inhibitor of the steroid dehydrogenase enzymes 11 β-hydroxysteroid dehydrogenase (human) and 3α, 20 β-hydroxysteroid dehydrogenase (bacterial). It is used for the treatment of digestive tract ulcers, especially in the stomach. Used in the treatment of peptic, mainly gastric, ulcers. It is often used as a gap junction inhibitor in neuroscience research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water to 100 mM.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat.
TRUSTED_SUSTAINABILITY
Color White to Cream
Quantity 1 g
Merck Index 14,1793
Solubility Information Soluble in water to 100 mM.
Formula Weight 614.7
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material Carbenoxolone disodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'