Learn More
Paragon Scientific™ Cannon-Fenske Routine Viscometer for Transparent Liquids
For the determination of kinematic viscosity of transparent Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
Brand:  Paragon Scientific™ CFR09NC
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 16010000
  • 1656.06 NOK / Each
Estimated Shipment: 12-01-2022
Add to basket
Description
Description
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Specifications
D 445, D 446
ASTM D 445, D 446 and ISO 3104, 3105
No
1.2
400
240 to 1,200 cSt
Special Offers
4 week lead-time