Learn More
Paragon Scientific™ Cannon-Fenske Routine Viscometer for Transparent Liquids
Click to view available options
Viscosity Range:
0.5 to 2 cSt
0.8 to 4 cSt
1.6 to 8 cSt
10,000 to 50,000 cSt
100 to 500 cSt
1600 to 8000 cSt
20 to 100 cSt
20,000 to 100,000 cSt
240 to 1200 cSt
3 to 15 cSt
4000 to 20,000 cSt
50 to 250 cSt
500 to 2500 cSt
7 to 35 cSt
Unit Size:
Each
Description
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Specifications
| Viscosity Range | 20,000 to 100,000 cSt |
| Calibrated | No |
| Approximate Constant | 100 |
| ASTM Numbers | D 445, D 446 |
| Certifications/Compliance | ASTM D 445, D 446 and ISO 3104, 3105 |
| Size | 700 |
4 week lead-time