Learn More
Calcium Sulfate Hemihydrate, Honeywell Fluka™
Catalog Number p-7071777
Click to view available options
Quantity:
1 kg
25 kg
10034-76-1
Ca2H2O9S2
290.283
MFCD00799067
ZOMBKNNSYQHRCA-UHFFFAOYSA-J
calcium sulfate hemihydrate, gypsum hemihydrate, calcium sulphate hemihydrate, unii-3rw091j48v, crystacal, densite, hemihydrate gypsum, densite gypsum, gipshalbhydrat, p 23 sulfate
3033839
CHEBI:32584
dicalcium;disulfate;hydrate
O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ca+2].[Ca+2]
10034-76-1
Ca2H2O9S2
290.283
MFCD00799067
ZOMBKNNSYQHRCA-UHFFFAOYSA-J
calcium sulfate hemihydrate, gypsum hemihydrate, calcium sulphate hemihydrate, unii-3rw091j48v, crystacal, densite, hemihydrate gypsum, densite gypsum, gipshalbhydrat, p 23 sulfate
3033839
CHEBI:32584
dicalcium;disulfate;hydrate
O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ca+2].[Ca+2]
Purum,≥97.0%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Quantity 1 kg
Linear Formula CaSO4 · 0.5H2O
UN Number NONH for all modes of transport
Formula Weight 145.15g/mol
Percent Purity ≥97%
Grade purum
Chemical Name or Material Calcium Sulfate Hemihydrate