missing translation for 'onlineSavingsMsg'
Learn More

Caffeine monohydrate, MP Biomedicals™

Catalog Number p-4404322
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy
58-08-2
C8H10N4O2
194.19
MFCD00005758
RYYVLZVUVIJVGH-UHFFFAOYSA-N
caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
2519
CHEBI:27732
1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
CN1C=NC2=C1C(=O)N(C)C(=O)N2C
This item is not returnable. View return policy
58-08-2
C8H10N4O2
194.19
MFCD00005758
RYYVLZVUVIJVGH-UHFFFAOYSA-N
caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
2519
CHEBI:27732
1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
CN1C=NC2=C1C(=O)N(C)C(=O)N2C
Color White
Density 1.23g/mL
Assay Percent Range >99%
Quantity 500 g
Formula Weight 212.2
Physical Form Powder
Chemical Name or Material Caffeine monohydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'